missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human REXO2 Protein
A cDNA sequence encoding the REXO2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13303-20ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
REXO2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSVREGGAA MAAGESMAQR MVWVDLEMTG LDIEKDQIIE MACLITDSDL NILAEGPNLI IKQPDELLDS MSDWCKEHHG KSGLTKAVKE STITLQQAEY EFLSFVRQQT PPGLCPLAGN SVHEDKKFLD KYMPQFMKHL HYRIIDVSTV KELCRRWYPE EYEFAPKKAA SHRALDDISE SIKELQFYRN NIFKKKIDEK KRKIIENGEN EKTVS | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The REXO2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 20% glycerol. |