missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human REXO2 Protein
A cDNA sequence encoding the REXO2 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 3905.00 CHF
Spezifikation
Name | REXO2 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15983139
|
enQuireBio™
QP13303-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15973139
|
enQuireBio™
QP13303-20UG |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15963139
|
enQuireBio™
QP13303-1MG |
1 mg |
3905.00 CHF
1 Milligramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
REXO2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The REXO2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSVREGGAA MAAGESMAQR MVWVDLEMTG LDIEKDQIIE MACLITDSDL NILAEGPNLI IKQPDELLDS MSDWCKEHHG KSGLTKAVKE STITLQQAEY EFLSFVRQQT PPGLCPLAGN SVHEDKKFLD KYMPQFMKHL HYRIIDVSTV KELCRRWYPE EYEFAPKKAA SHRALDDISE SIKELQFYRN NIFKKKIDEK KRKIIENGEN EKTVS | |
Greater than 90% as determined by SDS-PAGE. |