missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RDBP Protein
A cDNA sequence encoding the RDBP was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13291-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
7936 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MLVIPPGLSE EEEALQKKFN KLKKKKKALL ALKKQSSSST TSQGGVKRSL SEQPVMDTAT ATEQAKQLVK SGAISAIKAE TKNSGFKRSR TLEGKLKDPE KGPVPTFQPF QRSISADDDL QESSRRPQRK SLYESFVSSS DRLRELGPDG EEAEGPGAGD GPPRSFDWGY EERSGAHSSA SPPRSRSRDR SHERNRDRDR DRERDRDRDR DRDRERDRDR DRDRDRDRER DRDRERDRDR DREGPFRRSD SFPERRAPRK GNTLYVYGED MTPTLLRGAF SPFGNIIDLS MDPPRNCAFV TYEKMESADQ AVAELNGTQV ESVQLKVNIA RKQPMLDAAT GKSVWGSLAV QNSPKGCHRD KRTQIVYSDD VYKENLVDGF | |
RDBP purity was found to be greater than 85% as determined by SDS-PAGE. |
RDBP Protein | |
Research Use Only | |
NELFE | |
Human | |
His | |
The RDBP protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 2mM DTT and 10% glycerol. |