missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAET1G Protein
A cDNA sequence encoding the RAET1G was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13255-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
RAET1G Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASGLADPHSLCY DITVIPKFRP GPRWCAVQGQ VDEKTFLHYD CGSKTVTPVS PLGKKLNVTT AWKAQNPVLR EVVDILTEQL LDIQLENYIP KEPLTLQARM SCEQKAEGHG SGSWQLSFDG QIFLLFDSEN RMWTTVHPGA RKMKEKWEND KDMTMSFHYI SMGDCTGWLE DFLMGMDSTL EPSAGGTV | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RAET1G filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 20mM Tris buffer and 50mM NaCl, pH 7.5. |