missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB7A Protein
A cDNA sequence encoding the RAB7A was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13246-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
RAB7A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MTSRKKVLLK VIILGDSGVG KTSLMNQYVN KKFSNQYKAT IGADFLTKEV MVDDRLVTMQ IWDTAGQERF QSLGVAFYRG ADCCVLVFDV TAPNTFKTLD SWRDEFLIQA SPRDPENFPF VVLGNKIDLE NRQVATKRAQ AWCYSKNNIP YFETSAKEAINVEQAFQTIA RNALKQETEV ELYNEFPEPI KLDKNDRAKA SAESCSC | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RAB7A protein solution (0.5 mg/ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 100mM NaCl and 30% glycerol. |