missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTRH2 Protein
A cDNA sequence encoding the PTRH2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13214-20ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
51651 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEYKMILVVR NDLKMGKGKV AAQCSHAAVS AYKQIQRRNP EMLKQWEYCG QPKVVVKAPD EETLIALLAH AKMLGLTVSL IQDAGRTQIA PGSQTVLGIG PGPADLIDKV TGHLKLY | |
Greater than 95.0% as determined by SDS-PAGE. |
PTRH2 Protein | |
Research Use Only | |
PTRH2 | |
Human | |
His | |
The PTRH2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 1mM DTT. |