missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTRH2 Protein
A cDNA sequence encoding the PTRH2 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 5210.00 CHF
Spezifikation
Gen-ID (Entrez) | 51651 |
---|---|
Name | PTRH2 Protein |
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gensymbol | PTRH2 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15902869
|
enQuireBio™
QP13214-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15992859
|
enQuireBio™
QP13214-20UG |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15982859
|
enQuireBio™
QP13214-1MG |
1 mg |
5210.00 CHF
1 Milligramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
51651 | |
Research Use Only | |
PTRH2 | |
Human | |
His | |
The PTRH2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 1mM DTT. |
PTRH2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEYKMILVVR NDLKMGKGKV AAQCSHAAVS AYKQIQRRNP EMLKQWEYCG QPKVVVKAPD EETLIALLAH AKMLGLTVSL IQDAGRTQIA PGSQTVLGIG PGPADLIDKV TGHLKLY | |
Greater than 95.0% as determined by SDS-PAGE. |