missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSMD13 Protein
A cDNA sequence encoding the PSMD13 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13192-20ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
PSMD13 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMKDVPGF LQQSQNSGPG QPAVWHRLEE LYTKKLWHQL TLQVLDFVQD PCFAQGDGLI KLYENFISEF EHRVNPLSLV EIILHVVRQM TDPNVALTFL EKTREKVKSS DEAVILCKTA IGALKLNIGD LQVTKETIED VEEMLNNLPG VTSVHSRFYD LSSKYYQTIG NHASYYKDAL RFLGCVDIKD LPVSEQQERA FTLGLAGLLG EGVFNFGELL MHPVLESLRN TDRQWLIDTL YAFNSGNVER FQTLKTAWGQ QPDLAANEAQ LLRKIQLLCL MEMTFTRPAN HRQLTFEEIA KSAKITVNEV ELLVMKALSV GLVKGSIDEV DKRVHMTWVQ PRVLDLQQIK GMKDRLEFWC TDVKSMEMLV EHQAHDILT | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PSMD13 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |