missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PSAT1 Protein

A cDNA sequence encoding the PSAT1 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13178-20ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15972739

  • 203.00 CHF / 20 Mikrogramm
Verfügbar ab: 19-09-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

PSAT1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMDAPRQ VVNFGPGPAK LPHSVLLEIQ KELLDYKGVG ISVLEMSHRS SDFAKIINNT ENLVRELLAV PDNYKVIFLQ GGGCGQFSAV PLNLIGLKAG RCADYVVTGA WSAKAAEEAK KFGTINIVHP KLGSYTKIPD PSTWNLNPDA SYVYYCANET VHGVEFDFIP DVKGAVLVCD MSSNFLSKPV DVSKFGVIFA GAQKNVGSAG VTVVIVRDDL LGFALRECPS VLEYKVQAGN SSLYNTPPCF SIYVMGLVLE WIKNNGGAAA MEKLSSIKSQ TIYEIIDNSQ GFYVCPVEPQ NRSKMNIPFR IGNAKGDDAL EKRFLDKALE LNMLSLKGHR SVGGIRASLY NAVTIEDVQK LAAFMKKFLE MHQL
Greater than 90.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The PSAT1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol and 1mM DTT.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Special Offers

Special Offers