missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSAT1 Protein
A cDNA sequence encoding the PSAT1 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 3905.00 CHF
Spezifikation
Name | PSAT1 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15982739
|
enQuireBio™
QP13178-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15972739
|
enQuireBio™
QP13178-20UG |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15962739
|
enQuireBio™
QP13178-1MG |
1 mg |
3905.00 CHF
1 Milligramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
PSAT1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PSAT1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMDAPRQ VVNFGPGPAK LPHSVLLEIQ KELLDYKGVG ISVLEMSHRS SDFAKIINNT ENLVRELLAV PDNYKVIFLQ GGGCGQFSAV PLNLIGLKAG RCADYVVTGA WSAKAAEEAK KFGTINIVHP KLGSYTKIPD PSTWNLNPDA SYVYYCANET VHGVEFDFIP DVKGAVLVCD MSSNFLSKPV DVSKFGVIFA GAQKNVGSAG VTVVIVRDDL LGFALRECPS VLEYKVQAGN SSLYNTPPCF SIYVMGLVLE WIKNNGGAAA MEKLSSIKSQ TIYEIIDNSQ GFYVCPVEPQ NRSKMNIPFR IGNAKGDDAL EKRFLDKALE LNMLSLKGHR SVGGIRASLY NAVTIEDVQK LAAFMKKFLE MHQL | |
Greater than 90.0% as determined by SDS-PAGE. |