missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRKAG1 Protein
A cDNA sequence encoding the PRKAG1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13142-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
PRKAG1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMETVISS DSSPAVENEH PQETPESNNS VYTSFMKSHR CYDLIPTSSK LVVFDTSLQV KKAFFALVTN GVRAAPLWDS KKQSFVGMLT ITDFINILHR YYKSALVQIY ELEEHKIETW REVYLQDSFK PLVCISPNAS LFDAVSSLIR NKIHRLPVID PESGNTLYIL THKRILKFLK LFITEFPKPE FMSKSLEELQ IGTYANIAMV RTTTPVYVAL GIFVQHRVSA LPVVDEKGRV VDIYSKFDVI NLAAEKTYNN LDVSVTKALQ HRSHYFEGVL KCYLHETLET IINRLVEAEV HRLVVVDEND VVKGIVSLSD ILQALVLTGG EKKP | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PRKAG1 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |