missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRKAG1 Protein
A cDNA sequence encoding the PRKAG1 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 7525.00 CHF
Spezifikation
Name | PRKAG1 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15902639
|
enQuireBio™
QP13142-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15982629
|
enQuireBio™
QP13142-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15992629
|
enQuireBio™
QP13142-1MG |
1 mg |
7525.00 CHF
1 Milligramm |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
PRKAG1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PRKAG1 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMETVISS DSSPAVENEH PQETPESNNS VYTSFMKSHR CYDLIPTSSK LVVFDTSLQV KKAFFALVTN GVRAAPLWDS KKQSFVGMLT ITDFINILHR YYKSALVQIY ELEEHKIETW REVYLQDSFK PLVCISPNAS LFDAVSSLIR NKIHRLPVID PESGNTLYIL THKRILKFLK LFITEFPKPE FMSKSLEELQ IGTYANIAMV RTTTPVYVAL GIFVQHRVSA LPVVDEKGRV VDIYSKFDVI NLAAEKTYNN LDVSVTKALQ HRSHYFEGVL KCYLHETLET IINRLVEAEV HRLVVVDEND VVKGIVSLSD ILQALVLTGG EKKP | |
Greater than 85.0% as determined by SDS-PAGE. |