missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PRKAG1 Protein

A cDNA sequence encoding the PRKAG1 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13142-2ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15902639

  • 131.00 CHF / 2 Mikrogramm
Verfügbar ab: 28-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

PRKAG1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMETVISS DSSPAVENEH PQETPESNNS VYTSFMKSHR CYDLIPTSSK LVVFDTSLQV KKAFFALVTN GVRAAPLWDS KKQSFVGMLT ITDFINILHR YYKSALVQIY ELEEHKIETW REVYLQDSFK PLVCISPNAS LFDAVSSLIR NKIHRLPVID PESGNTLYIL THKRILKFLK LFITEFPKPE FMSKSLEELQ IGTYANIAMV RTTTPVYVAL GIFVQHRVSA LPVVDEKGRV VDIYSKFDVI NLAAEKTYNN LDVSVTKALQ HRSHYFEGVL KCYLHETLET IINRLVEAEV HRLVVVDEND VVKGIVSLSD ILQALVLTGG EKKP
Greater than 85.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The PRKAG1 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote