missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POMZP3 Protein
A cDNA sequence encoding the POMZP3 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13109-20ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
POMZP3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMVCSPVT LRIAPPDRRF SRSAIPEQII SSTLSSPSSN APDPCAKETV LSALKEKKKK RTVEEEDQIF LDGQENKRSC LVDGLTDASS AFKVPRPGPD TLQFTVDVFH FANDSRNMIY ITCHLKVTLA EQDPDELNKA CSFSKPSNSW FPVEGLADIC QCCNKGDCGT PSHSRRQPRV VSQWSTSASL | |
Greater than 85% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The POMZP3 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |