missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human POLR2K Protein

A cDNA sequence encoding the POLR2K was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13105-2ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15982519

  • 131.00 CHF / 2 Mikrogramm
Verfügbar ab: 27-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

POLR2K Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMDTQKDV QPPKQQPMIY ICGECHTENE IKSRDPIRCR ECGYRIMYKK RTKRLVVFDA R
Greater than 85.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
POLR2K protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote