missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POLR2K Protein
A cDNA sequence encoding the POLR2K was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 7525.00 CHF
Spezifikation
Name | POLR2K Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15982519
|
enQuireBio™
QP13105-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15962519
|
enQuireBio™
QP13105-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15972519
|
enQuireBio™
QP13105-1MG |
1 mg |
7525.00 CHF
1 Milligramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
POLR2K Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
POLR2K protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDTQKDV QPPKQQPMIY ICGECHTENE IKSRDPIRCR ECGYRIMYKK RTKRLVVFDA R | |
Greater than 85.0% as determined by SDS-PAGE. |