missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human POLD4 Protein

A cDNA sequence encoding the POLD4 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13092-2ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15992479

  • 131.00 CHF / 2 Mikrogramm
Verfügbar ab: 01-07-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

57804
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMGRKRLI TDSYPVVKRR EGPAGHSKGE LAPELGEEPQ PRDEEEAELE LLRQFDLAWQ YGPCTGITRL QRWCRAKQMG LEPPPEVWQV LKTHPGDPRF QCSLWHLYPL
Greater than 90.0% as determined by SDS-PAGE.
POLD4 Protein
Research Use Only
POLD4
Human
His
The POLD4 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 40% glycerol and 2mM DTT.
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali