missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PKAR-I alpha Protein

A cDNA sequence encoding the PKAR-I alpha was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP13062-50ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15952389

  • 2230.00 CHF / 50 Mikrogramm
Verfügbar ab: 25-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

5573
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MESGSTAASE EARSLRECEL YVQKHNIQAL LKDSIVQLCT ARPERPMAFL REYFERLEKE EAKQIQNLQK AGTRTDSRED EISPPPPNPV VKGRRRRGAI SAEVYTEEDA ASYVRKVIPK DYKTMAALAK AIEKNVLFSH LDDNERSDIF DAMFSVSFIA GETVIQQGDE GDNFYVIDQG ETDVYVNNEW ATSVGEGGSF GELALIYGTP RAATVKAKTN VKLWGIDRDS YRRILMGSTL RKRKMYEEFL SKVSILESLD KWERLTVADA LEPVQFEDGQ KIVVQGEPGD EFFIILEGSA AVLQRRSENE EFVEVGRLGP SDYFGEIALL MNRPRAATVV ARGPLKCVKL DRPRFERVLG PCSDILKRNI QQYNSFVSLS V
Greater than 95% as determined by SDS-PAGE.
PKAR-I alpha Protein
Research Use Only
PRKAR1A
Human
Untagged
PKA regulatory subunit-I alpha is supplied at a concentration of 1.3 mg/ml in 20mM MOPS (pH 7.0), 150mM NaCl, 1mM 2-mercaptoethanol and 50% glycerol.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote