missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PET117 Protein
A cDNA sequence encoding the PET117 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13011-2ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
PET117 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSVHVKQQW DQQRLRDGVI RDIERQIRKK ENIRLLGEQI ILTEQLEAER EKMLLAKGSQ KS | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PET117 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol and 2mM DTT. |