missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDGF AB Protein
A cDNA sequence encoding the PDGF AB was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10804-2ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
PDGF AB Protein | |
Research Use Only | |
The ED50 as determined by the dose-dependent proliferation of mouse 3T3 indicator cells, is 1.4-2.1 ng/ml. This corresponds to a specific activity of 7.1x 105 units/mg. | |
Human | |
Untagged | |
Lyophilized from 10mM AcOH (Acetic Acid). |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
Alpha chain: MSIEEAVPAV CKTRTVIYEI PRSQVDPTSA NFLIWPPCVE VKRCTGCCNT SSVKCQPSRV HHRSVKVAKV EYVRKKPKLK EVQVRLEEHL ECACATTSLN PDYREEDTGR PRESGKKRKR KRLKPT Beta chain: SLGSLTIAEP AMIAECKTRT EVFEISRRLI DRTNANFLVW PPCVEVQRCS GCCNNRNVQC RPTQVQLRPV QVRKIGIVRK KPIFKKATVT LGDHLACKCE TVAAARPVT | |
Greater than 95.0% as determined by SDS-PAGE analysis. |