missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDGF AB Protein
A cDNA sequence encoding the PDGF AB was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 5090.00 CHF
Spezifikation
Name | PDGF AB Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biologische Aktivität | The ED50 as determined by the dose-dependent proliferation of mouse 3T3 indicator cells, is 1.4-2.1 ng/ml. This corresponds to a specific activity of 7.1x 105 units/mg. |
Produkttyp | Recombinant Protein |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15985218
|
enQuireBio™
QP10804-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 19-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15965218
|
enQuireBio™
QP10804-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 19-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15975218
|
enQuireBio™
QP10804-1MG |
1 mg |
5090.00 CHF
1 Milligramm |
Verfügbar ab: 19-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
PDGF AB Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
Alpha chain: MSIEEAVPAV CKTRTVIYEI PRSQVDPTSA NFLIWPPCVE VKRCTGCCNT SSVKCQPSRV HHRSVKVAKV EYVRKKPKLK EVQVRLEEHL ECACATTSLN PDYREEDTGR PRESGKKRKR KRLKPT Beta chain: SLGSLTIAEP AMIAECKTRT EVFEISRRLI DRTNANFLVW PPCVEVQRCS GCCNNRNVQC RPTQVQLRPV QVRKIGIVRK KPIFKKATVT LGDHLACKCE TVAAARPVT | |
Greater than 95.0% as determined by SDS-PAGE analysis. |
Research Use Only | |
The ED50 as determined by the dose-dependent proliferation of mouse 3T3 indicator cells, is 1.4-2.1 ng/ml. This corresponds to a specific activity of 7.1x 105 units/mg. | |
Human | |
Untagged | |
Lyophilized from 10mM AcOH (Acetic Acid). |