missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PDGF AB Protein

A cDNA sequence encoding the PDGF AB was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP10804-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15975218

  • 5090.00 CHF / 1 Milligramm
Verfügbar ab: 28-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

PDGF AB Protein
Research Use Only
The ED50 as determined by the dose-dependent proliferation of mouse 3T3 indicator cells, is 1.4-2.1 ng/ml. This corresponds to a specific activity of 7.1x 105 units/mg.
Human
Untagged
Lyophilized from 10mM AcOH (Acetic Acid).
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
Alpha chain: MSIEEAVPAV CKTRTVIYEI PRSQVDPTSA NFLIWPPCVE VKRCTGCCNT SSVKCQPSRV HHRSVKVAKV EYVRKKPKLK EVQVRLEEHL ECACATTSLN PDYREEDTGR PRESGKKRKR KRLKPT Beta chain: SLGSLTIAEP AMIAECKTRT EVFEISRRLI DRTNANFLVW PPCVEVQRCS GCCNNRNVQC RPTQVQLRPV QVRKIGIVRK KPIFKKATVT LGDHLACKCE TVAAARPVT
Greater than 95.0% as determined by SDS-PAGE analysis.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote