missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human p21/CIP1/CDKN1A His Protein
Alle ansehen Bio Techne Produkte
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikrogramm
Beschreibung
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-164 of Human p21/CIP1/CDKN1A The Recombinant Human p21/CIP1/CDKN1A Protein is derived from E. coli. The Recombinant Human p21/CIP1/CDKN1A Protein has been validated for the following applications: SDS-Page.
Spezifikation
Spezifikation
| Konzentration | 0.5mg/mL |
| Zur Verwendung mit (Anwendung) | SDS-PAGE |
| Zusammensetzung | 20mM Tris-HCl buffer (pH 8.0) containing 2M urea, 10% glycerol, 0.1M NaCl |
| Gen-ID (Entrez) | 1026 |
| Molekulargewicht | 20.2 kDa |
| Menge | 100 μg |
| Quelle | E. coli |
| Lagerungsbedingungen | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Gensymbol | CDKN1A |
| Spezies | Human |
| Mehr anzeigen |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction