missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Osteocrin Protein

A cDNA sequence encoding the Osteocrin was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12935-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15902009

  • 5645.00 CHF / 1 Milligramm
Verfügbar ab: 01-07-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

Osteocrin Protein
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASHMVDVT TTEAFDSGVI DVQSTPTVRE EKSATDLTAK LLLLDELVSL ENDVIETKKK RSFSGFGSPL DRLSAGSVDH KGKQRKVVDH PKRRFGIPMD RIGRNRLSNS RG
Greater than 95% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer, pH 4.0.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote