missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NUBP1 Protein
A cDNA sequence encoding the NUBP1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12894-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
NUBP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEEVPHD CPGADSAQAG RGASCQGCPN QRLCASGAGA TPDTAIEEIK EKMKTVKHKI LVLSGKGGVG KSTFSAHLAH GLAEDENTQI ALLDIDICGP SIPKIMGLEG EQVHQSGSGW SPVYVEDNLG VMSVGFLLSS PDDAVIWRGP KKNGMIKQFL RDVDWGEVDY LIVDTPPGTS DEHLSVVRYL ATAHIDGAVI ITTPQEVSLQ DVRKEINFCR KVKLPIIGVV ENMSGFICPK CKKESQIFPP TTGGAELMCQ DLEVPLLGRV PLDPLIGKNC DKGQSFFIDA PDSPATLAYR SIIQRIQEFC NLHQSKEENL ISS | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NUBP1 protein solution (1 mg/ml) in Phosphate buffered saline (pH 7.4),10% glycerol and 1mM DTT. |