missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NNT1 Protein
A cDNA sequence encoding the NNT1 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 6775.00 CHF
Spezifikation
Gen-ID (Entrez) | 23529 |
---|---|
Name | NNT1 Protein |
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gensymbol | CLCF1 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15985188
|
enQuireBio™
QP10794-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab:
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15965188
|
enQuireBio™
QP10794-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab:
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15975188
|
enQuireBio™
QP10794-1MG |
1 mg |
6775.00 CHF
1 Milligramm |
Verfügbar ab:
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
23529 | |
Research Use Only | |
CLCF1 | |
Recombinant Protein | |
E. coli | |
MLNRTGDPGP GPSIQKTYDL TRYLEHQLRS LAGTYLNYLG PPFNEPDFNP PRLGAETLPR ATVDLEVWRS LNDKLRLTQN YEAYSHLLCY LRGLNRQAAT AELRRSLAHF CTSLQGLLGS IAGVMAALGY PLPQPLPGTE PTWTPGPAHS DFLQKMDDFW LLKELQTWLW RSAKDFNRLK KKMQPPAAAV TLHLGAHGF | |
Greater than 95.0% as determined by SDS-PAGE. |
NNT1 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50, as measured in a cell proliferation assay using human TF?1 cells transfected with human CNTF R?, is less than 15ng/ml. | |
Human | |
Untagged | |
NNT-1 protein was lyophilized from a 0.2microm filtered solution in Acetonitrile and TFA. |