missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NNT1 Protein
A cDNA sequence encoding the NNT1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10794-2ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
23529 | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50, as measured in a cell proliferation assay using human TF?1 cells transfected with human CNTF R?, is less than 15ng/ml. | |
Human | |
Untagged | |
NNT-1 protein was lyophilized from a 0.2microm filtered solution in Acetonitrile and TFA. |
NNT1 Protein | |
Research Use Only | |
CLCF1 | |
Recombinant Protein | |
E. coli | |
MLNRTGDPGP GPSIQKTYDL TRYLEHQLRS LAGTYLNYLG PPFNEPDFNP PRLGAETLPR ATVDLEVWRS LNDKLRLTQN YEAYSHLLCY LRGLNRQAAT AELRRSLAHF CTSLQGLLGS IAGVMAALGY PLPQPLPGTE PTWTPGPAHS DFLQKMDDFW LLKELQTWLW RSAKDFNRLK KKMQPPAAAV TLHLGAHGF | |
Greater than 95.0% as determined by SDS-PAGE. |