missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NFE2L2 Protein

A cDNA sequence encoding the NFE2L2 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12834-10ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15911699

  • 203.00 CHF / 10 Mikrogramm
Verfügbar ab: 25-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

4780
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MMDLELPPPG LPSQQDMDLI DILWRQDIDL GVSREVFDFS QRRKEYELEK QKKLEKERQE QLQKEQEKAF FAQLQLDEET GEFLPIQPAQ HIQSETSGSA NYSQVAHIPK SDALYFDDCM QLLAQTFPFV DDNEVSSATF QSLVPDIPGH IESPVFIATN QAQSPETSVA QVAPVDLDGM QQDIEQVWEE LLSIPELQCL NIENDKLVET TMVPSPEAKL TEVDNYHFYS SIPSMEKEVG NCSPHFLNAF EDSFSSILST EDPNQLTVNS LNSDATVNTD FGDEFYSAFI AEPSISNSMP SPATLSHSLS ELLNGPIDVS DLSLCKAFNQ NHPESTAEFN DSDSGISLNT SPSVASPEHS VESSSYGDTL LGLSDSEVEE LDSAPGSVKQ NGPKTPVHSS GDMVQPLSPS QGQSTHVHDA QCENTPEKEL PVSPGHRKTP FTKDKHSSRL EAHLTRDELR AKALHIPFPV EKIINLPVVD FNEMMSKEQF NEAQLALIRD IRRRGKNKVA AQNCRKRKLE NIVELEQDLD HLKDEKEKLL KEKGENDKSL HLLKKQLSTL YLEVFSMLRD EDGKPYSPSE YSLQQTRDGN VFLVPKSKKP DVKKN
Greater than 85.0% as determined by SDS-PAGE.
NFE2L2 Protein
Research Use Only
NFE2L2
Human
His
NFE2L2 protein solution (0.25 mg/ml) in Phosphate buffer saline (pH 7.4) containing 10% glycerol.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote