missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NFE2L2 Protein
A cDNA sequence encoding the NFE2L2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12834-100ug
Dettagli aggiuntivi : Gewicht : 0.01000kg
Specifica
4780 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MMDLELPPPG LPSQQDMDLI DILWRQDIDL GVSREVFDFS QRRKEYELEK QKKLEKERQE QLQKEQEKAF FAQLQLDEET GEFLPIQPAQ HIQSETSGSA NYSQVAHIPK SDALYFDDCM QLLAQTFPFV DDNEVSSATF QSLVPDIPGH IESPVFIATN QAQSPETSVA QVAPVDLDGM QQDIEQVWEE LLSIPELQCL NIENDKLVET TMVPSPEAKL TEVDNYHFYS SIPSMEKEVG NCSPHFLNAF EDSFSSILST EDPNQLTVNS LNSDATVNTD FGDEFYSAFI AEPSISNSMP SPATLSHSLS ELLNGPIDVS DLSLCKAFNQ NHPESTAEFN DSDSGISLNT SPSVASPEHS VESSSYGDTL LGLSDSEVEE LDSAPGSVKQ NGPKTPVHSS GDMVQPLSPS QGQSTHVHDA QCENTPEKEL PVSPGHRKTP FTKDKHSSRL EAHLTRDELR AKALHIPFPV EKIINLPVVD FNEMMSKEQF NEAQLALIRD IRRRGKNKVA AQNCRKRKLE NIVELEQDLD HLKDEKEKLL KEKGENDKSL HLLKKQLSTL YLEVFSMLRD EDGKPYSPSE YSLQQTRDGN VFLVPKSKKP DVKKN | |
Greater than 85.0% as determined by SDS-PAGE. |
NFE2L2 Protein | |
Research Use Only | |
NFE2L2 | |
Human | |
His | |
NFE2L2 protein solution (0.25 mg/ml) in Phosphate buffer saline (pH 7.4) containing 10% glycerol. |