missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NAP1L4 Protein
A cDNA sequence encoding the NAP1L4 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12792-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
NAP1L4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMADHSFS DGVPSDSVEA AKNASNTEKL TDQVMQNPRV LAALQERLDN VPHTPSSYIE TLPKAVKRRI NALKQLQVRC AHIEAKFYEE VHDLERKYAA LYQPLFDKRR EFITGDVEPT DAESEWHSEN EEEEKLAGDM KSKVVVTEKA AATAEEPDPK GIPEFWFTIF RNVDMLSELV QEYDEPILKH LQDIKVKFSD PGQPMSFVLE FHFEPNDYFT NSVLTKTYKM KSEPDKADPF SFEGPEIVDC DGCTIDWKKG KNVTVKTIKK KQKHKGRGTV RTITKQVPNE SFFNFFNPLK ASGDGESLDE DSEFTLASDF EIGHFFRERI VPRAVLYFTG EAIEDDDNFE EGEEGEEEEL EGDEEGEDED DAEINPKV | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NAP1L4 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |