missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NAP1L4 Protein
A cDNA sequence encoding the NAP1L4 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 3905.00 CHF
Spezifikation
Name | NAP1L4 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15911559
|
enQuireBio™
QP12792-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15901559
|
enQuireBio™
QP12792-20UG |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15991549
|
enQuireBio™
QP12792-1MG |
1 mg |
3905.00 CHF
1 Milligramm |
Verfügbar ab: 21-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
NAP1L4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NAP1L4 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMADHSFS DGVPSDSVEA AKNASNTEKL TDQVMQNPRV LAALQERLDN VPHTPSSYIE TLPKAVKRRI NALKQLQVRC AHIEAKFYEE VHDLERKYAA LYQPLFDKRR EFITGDVEPT DAESEWHSEN EEEEKLAGDM KSKVVVTEKA AATAEEPDPK GIPEFWFTIF RNVDMLSELV QEYDEPILKH LQDIKVKFSD PGQPMSFVLE FHFEPNDYFT NSVLTKTYKM KSEPDKADPF SFEGPEIVDC DGCTIDWKKG KNVTVKTIKK KQKHKGRGTV RTITKQVPNE SFFNFFNPLK ASGDGESLDE DSEFTLASDF EIGHFFRERI VPRAVLYFTG EAIEDDDNFE EGEEGEEEEL EGDEEGEDED DAEINPKV | |
Greater than 90.0% as determined by SDS-PAGE. |