missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MRM1 Protein
A cDNA sequence encoding the MRM1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12728-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
MRM1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSHAARHG ERPGGEELSR LLLDDLVPTS RLELLFGMTP CLLALQAARR SVARLLLQAG KAGLQGKRAE LLRMAEARDI PVLRPRRQKL DTMCRYQVHQ GVCMEVSPLR PRPWREAGEA SPGDDPQQLW LVLDGIQDPR NFGAVLRSAH FLGVDKVITS RRNSCPLTPV VSKSSAGAME VMDVFSTDDL TGFLQTKAQQ GWLVAGTVGC PSTEDPQSSE IPIMSCLEFL WERPTLLVLG NEGSGLSQEV QASCQLLLTI LPRRQLPPGL ESLNVSVAAG ILLHSICSQR KGFPTEGERR QLLQDPQEPS ARSEGLSMAQ HPGLSSGPEK ERQNEG | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MRM1 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol, 1mM DTT and 0.1mM PMSF. |