missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MPO Protein
A cDNA sequence encoding the MPO was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 3620.00 CHF
Spezifikation
Name | MPO Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15931339
|
enQuireBio™
QP12722-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 25-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15921339
|
enQuireBio™
QP12722-25UG |
25 μg |
203.00 CHF
25 Mikrogramm |
Verfügbar ab: 25-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15911339
|
enQuireBio™
QP12722-1MG |
1 mg |
3620.00 CHF
1 Milligramm |
Verfügbar ab: 25-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
MPO Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
MPO solution is supplied in 20mM HEPES buffer pH 7.5, 200mM NaCl and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
Human | |
MGVPFFSSLR CMVDLGPCWA GGLTAEMKLL LALAGLLAIL ATPQPSEGAA PAVLGEVDTS LVLSSMEEAK QLVDKAYKER RESIKQRLRS GSASPMELLS YFKQPVAATR TAVRAADYLH VALDLLERKL RSLWRRPFNV TDVLTPAQLN VLSKSSGCAY QDVGVTCPEQ DKYRTITGMC NNRRSPTLGA SNRAFVRWLP AEYEDGFSLP YGWTPGVKRN GFPVALARAV SNEIVRFPTD QLTPDQERSL MFMQWGQLLD HDLDFTPEPA ARASFVTGVN CETSCVQQPP CFPLKIPPND PRIKNQADCI PFFRSCPACP GSNITIRNQI NALTSFVDAS MVYGSEEPLA RNLRNMSNQL GLLAVNQRFQ DNGRALLPFD NLHDDPCLLT NRSARIPCFL AGDTRSSEMP ELTSMHTLLL REHNRLATEL KSLNPRWDGE RLYQEARKIV GAMVQIITYR DYLPLVLGPT AMRKYLPTYR SYNDSVDPRI ANVFTNAFRY GHTLIQPFMF RLDNRYQPME PNPRVPLSRV FFASWRVVLE GGIDPILRGL MATPAKLNRQ NQIAVDEIRE RLFEQVMRIG LDLPALNMQR SRDHGLPGYN AWRRFCGLPQ PETVGQLGTV LRNLKLARKL MEQYGTPNNI DIWMGGVSEP LKRKGRVGPL LACIIGTQFR KLRDGDRFWW ENEGVFSMQQ RQALAQISLP RIICDNTGIT TVSKNNIFMS NSYPRDFVNC STLPALNLAS WREAS | |
Greater than 80% as determined by SDS-PAGE. |