missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MPO Protein
A cDNA sequence encoding the MPO was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12722-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
MPO Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
MGVPFFSSLR CMVDLGPCWA GGLTAEMKLL LALAGLLAIL ATPQPSEGAA PAVLGEVDTS LVLSSMEEAK QLVDKAYKER RESIKQRLRS GSASPMELLS YFKQPVAATR TAVRAADYLH VALDLLERKL RSLWRRPFNV TDVLTPAQLN VLSKSSGCAY QDVGVTCPEQ DKYRTITGMC NNRRSPTLGA SNRAFVRWLP AEYEDGFSLP YGWTPGVKRN GFPVALARAV SNEIVRFPTD QLTPDQERSL MFMQWGQLLD HDLDFTPEPA ARASFVTGVN CETSCVQQPP CFPLKIPPND PRIKNQADCI PFFRSCPACP GSNITIRNQI NALTSFVDAS MVYGSEEPLA RNLRNMSNQL GLLAVNQRFQ DNGRALLPFD NLHDDPCLLT NRSARIPCFL AGDTRSSEMP ELTSMHTLLL REHNRLATEL KSLNPRWDGE RLYQEARKIV GAMVQIITYR DYLPLVLGPT AMRKYLPTYR SYNDSVDPRI ANVFTNAFRY GHTLIQPFMF RLDNRYQPME PNPRVPLSRV FFASWRVVLE GGIDPILRGL MATPAKLNRQ NQIAVDEIRE RLFEQVMRIG LDLPALNMQR SRDHGLPGYN AWRRFCGLPQ PETVGQLGTV LRNLKLARKL MEQYGTPNNI DIWMGGVSEP LKRKGRVGPL LACIIGTQFR KLRDGDRFWW ENEGVFSMQQ RQALAQISLP RIICDNTGIT TVSKNNIFMS NSYPRDFVNC STLPALNLAS WREAS | |
Greater than 80% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
MPO solution is supplied in 20mM HEPES buffer pH 7.5, 200mM NaCl and 20% glycerol. |