missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MICA Protein
A DNA sequence encoding the Human MICA, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10171-ec-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_000238.1 | |
MICA Protein | |
Research Use Only | |
MICA | |
The specific activity is determined by binding MICA antibody in ELISA. | |
Human | |
His | |
0.2 um filtered 30 % Acetonitrile, 0.1% TFA, lyophilized |
100507436 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MSYYHHHHHH DYDIPTTENL YFQGAMAPEF EPHSLRYNLT VLSWDGSVQS GFLTEVHLDG QPFLRCDRQK CRAKPQGQWA EDVLGNKTWD RETRDLTGNG KDLRMTLAHI KDQKEGLHSL QEIRVCEIHE DNSTRSSQHF YYDGELFLSQ NLETKEWTMP QSSRAQTLAM NVRNFLKEDA MKTKTHYHAM HADCLQELRR YLKSGVVLRR TVPPMVNVTR SEASEGNITV TCRASGFYPW NITLSWRQDG VSLSHDTQQW GDVLPDGNGT YQTWVATRIC QGEEQRFTCY MEHSGNHSTH PVPSGKVLVL QSHKLGCFGG | |
Greater than 95% as determined by SDS-PAGE and HPLC. |