missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human M6PR Protein

A cDNA sequence encoding the M6PR was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12616-10ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15901009

  • 203.00 CHF / 10 Mikrogramm
Verfügbar ab: 27-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

M6PR Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSTEEKTCD LVGEKGKESE KELALVKRLK PLFNKSFEST VGQGSDTYIY IFRVCREAGN HTSGAGLVQI NKSNGKETVV GRLNETHIFN GSNWIMLIYK GGDEYDNHCG KEQRRAVVMI SCNRHTLADN FNPVSEERGK VQDCFYLFEM DSSLACSPEI SH
Greater than 85.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The M6PR solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers