missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human M6PR Protein
A cDNA sequence encoding the M6PR was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 7525.00 CHF
Spezifikation
Name | M6PR Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15921009
|
enQuireBio™
QP12616-2UG |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15901009
|
enQuireBio™
QP12616-10UG |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15911009
|
enQuireBio™
QP12616-1MG |
1 mg |
7525.00 CHF
1 Milligramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
M6PR Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The M6PR solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSTEEKTCD LVGEKGKESE KELALVKRLK PLFNKSFEST VGQGSDTYIY IFRVCREAGN HTSGAGLVQI NKSNGKETVV GRLNETHIFN GSNWIMLIYK GGDEYDNHCG KEQRRAVVMI SCNRHTLADN FNPVSEERGK VQDCFYLFEM DSSLACSPEI SH | |
Greater than 85.0% as determined by SDS-PAGE. |