missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LDL Protein
A cDNA sequence encoding the LDL was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12550-100mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
LDL Protein | |
Research Use Only | |
Recombinant Protein | |
Human |
100 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged |