missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LCMT1 Protein
A cDNA sequence encoding the LCMT1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12544-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
LCMT1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMATRQR ESSITSCCST SSCDADDEGV RGTCEDASLC KRFAVSIGYW HDPYIQHFVR LSKERKAPEI NRGYFARVHG VSQLIKAFLR KTECHCQIVN LGAGMDTTFW RLKDEDLLPS KYFEVDFPMI VTRKLHSIKC KPPLSSPILE LHSEDTLQMD GHILDSKRYA VIGADLRDLS ELEEKLKKCN MNTQLPTLLI AECVLVYMTP EQSANLLKWA ANSFERAMFI NYEQVNMGDR FGQIMIENLR RRQCDLAGVE TCKSLESQKE RLLSNGWETA SAVDMMELYN RLPRAEVSRI ESLEFLDEME LLEQLMRHYC LCWATKGGNE LGLKEITY | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The LCMT1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT and 0.15M NaCl. |