missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LAMP2 Protein
A cDNA sequence encoding the LAMP2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12530-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
LAMP2 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MVCFRLFPVP GSGLVLVCLV LGAVRSYALE LNLTDSENAT CLYAKWQMNF TVRYETTNKT YKTVTISDHG TVTYNGSICG DDQNGPKIAV QFGPGFSWIA NFTKAASTYS IDSVSFSYNT GDNTTFPDAE DKGILTVDEL LAIRIPLNDL FRCNSLSTLE KNDVVQHYWD VLVQAFVQNG TVSTNEFLCD KDKTSTVAPT IHTTVPSPTT TPTPKEKPEA GTYSVNNGND TCLLATMGLQ LNITQDKVAS VININPNTTH STGSCRSHTA LLRLNSSTIK YLDFVFAVKN ENRFYLKEVN ISMYLVNGSV FSIANNNLSY WDAPLGSSYM CNKEQTVSVS GAFQINTFDL RVQPFNVTQG KYSTAQDCSA DDDNFLVPIA VGAALAGVLI LVLLAYFIGL KHHHAGYEQF |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
LAMP2 (0.94 mg/ml) is supplied in 16mM HEPES buffer pH 8.0, 130mM NaCl and 20% Glycerol. |