missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL1F5 / IL36RN Protein
A DNA sequence encoding the Human IL1F5 / IL36RN, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10343-ec-25ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_036407.1 | |
IL1F5 / IL36RN Protein | |
Research Use Only | |
IL36RN | |
The ED50 as determined by inhibiting IL-36 beta induced IL-8 secretion by human preadipocytes is less than 1.0 ug/ml, corresponding to a specific activity of > 1000 IU/mg in the presence of 20 ng/ml of recombinant human IL-36 beta. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
26525 | |
25 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
VLSGALCFRM KDSALKVLYL HNNQLLAGGL HAGKVIKGEE ISVVPNRWLD ASLSPVILGV QGGSQCLSCG VGQEPTLTLE PVNIMELYLG AKESKSFTFY RRDMGLTSSF ESAAYPGWFL CTVPEADQPV RLTQLPENGG WNAPITDFYF QQCD | |
Greater than 95% as determined by SDS-PAGE and HPLC. |