missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-17AF Protein
Recombinant Protein
Marke: enQuireBio™ QP5200-500ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_002181.1 | |
IL-17AF Protein | |
Research Use Only | |
IL17 | |
Recombinant Protein | |
E. coli | |
IL-17A:MIV KAGITIPRNP GCPNSEDKNF PRTVMVNLNI HNRNTNTNPK RSSDYYNRST SPWNLHRNED PERYPSVIWE AKCRHLGCIN ADGNVDYHMN SVPIQQEILV LRREPPHCPN SFRLEKILVS VGCTCVTPIV HHVA IL-17F :MRKIPKVGH TFFQKPESCP PVPGGSMKLD IGIINENQRV SMSRNIESRS TSPWNYTVTW DPNRYPSEVV QAQCRNLGCI NAQGKEDISM NSVPIQQETL VVRRKHQGCS VSFQLEKVLV TVGCTCVTPV IHHVQ | |
Greater than 95% as determined by SDS-PAGE. |
3605 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Production of IL-6 from mouse 3T3 cells was significantly induced by less than 50 ng/ml of Human IL-17AF, or activity of at least 2.0 x 10^4 units/mg. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |