missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGF1 Isoform 2 Protein
A DNA sequence encoding the Human IGF1 Isoform 2, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10392-ec-5mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_000609.1 | |
IGF1 Isoform 2 Protein | |
Research Use Only | |
IGF1 | |
The ED50 as determined by the stimulation of protein synthesis using rat L6 myoblasts is less than 10 ng/ml, corresponding to a specific activity of > 1.0 M 105 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered 20 mM Tris-HCl, pH 8.0, 150 mM NaCl, with 0.02 % Tween-20, lyophilized |
3479 | |
5 mg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MFPAMPLSSL FVNGPRTLCG AELVDALQFV CGDRGFYFNK PTGYGSSSRR APQTGIVDEC CFRSCDLRRL EMYCAPLKPA KSA | |
Greater than 90% as determined by SDS-PAGE and HPLC. |