missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFN omega 1 / IFNW1 Protein
A DNA sequence encoding the Human IFN omega 1 / IFNW1, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10411-ec-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_002168.1 | |
IFN omega 1 / IFNW1 Protein | |
Research Use Only | |
IFNW1 | |
The ED50 as determined by a chemotaxis bioassay using human TF-1 cells is less than 0.01 ng/ml, corresponding to a specific activity of > 1.0 M 108 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
3467 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
CDLPQNHGLL SRNTLVLLHQ MRRISPFLCL KDRRDFRFPQ EMVKGSQLQK AHVMSVLHEM LQQIFSLFHT ERSSAAWNMT LLDQLHTGLH QQLQHLETCL LQVVGEGESA GAISSPALTL RRYFQGIRVY LKEKKYSDCA WEVVRMEIMK SLFLSTNMQE RLRSKDRDLG SS | |
Greater than 97% as determined by SDS-PAGE and HPLC. |