missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IFN a 1a Protein

A cDNA sequence encoding the IFN a 1a was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP10672-20ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15984598

  • 131.00 CHF / 20 Mikrogramm
Verfügbar ab: 25-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

IFN a 1a Protein
Research Use Only
The specific activity as determined in a viral resistance assay viral ressistance assay was found to be 100,000,000IU/ mg.
Human
Untagged
The IFN-a 1a protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4.
20 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
CDLPETHSLD NRRTLMLLAQ MSRISPSSCL MDRHDFGFPQ EEFDGNQFQK APAISVLHEL IQQIFNLFTT KDSSAAWDED LLDKFCTELY QQLNDLEACV MQEERVGETP LMNADSILAV KKYFRRITLY LTEKKYSPCA WEVVRAEIMR SLSLSTNLQE RLRRKE
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote