missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFN a 1a Protein
A cDNA sequence encoding the IFN a 1a was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10672-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
IFN a 1a Protein | |
Research Use Only | |
The specific activity as determined in a viral resistance assay viral ressistance assay was found to be 100,000,000IU/ mg. | |
Human | |
Untagged | |
The IFN-a 1a protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
CDLPETHSLD NRRTLMLLAQ MSRISPSSCL MDRHDFGFPQ EEFDGNQFQK APAISVLHEL IQQIFNLFTT KDSSAAWDED LLDKFCTELY QQLNDLEACV MQEERVGETP LMNADSILAV KKYFRRITLY LTEKKYSPCA WEVVRAEIMR SLSLSTNLQE RLRRKE | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |