Learn More
enQuireBio™ Recombinant Human HOXC11 Protein
A cDNA sequence encoding the HOXC11 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12297-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
HOXC11 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMFNSVNL GNFCSPSRKE RGADFGERGS CASNLYLPSC TYYMPEFSTV SSFLPQAPSR QISYPYSAQV PPVREVSYGL EPSGKWHHRN SYSSCYAAAD ELMHRECLPP STVTEILMKN EGSYGGHHHP SAPHATPAGF YSSVNKNSVL PQAFDRFFDN AYCGGGDPPA EPPCSGKGEA KGEPEAPPAS GLASRAEAGA EAEAEEENTN PSSSGSAHSV AKEPAKGAAP NAPRTRKKRC PYSKFQIREL EREFFFNVYI NKEKRLQLSR MLNLTDRQVK IWFQNRRMKE KKLSRDRLQY FSGNPLL | |
Greater than 80.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
HOXC11 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |