missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Follistatin / FST Protein
Recombinant Protein
Marke: enQuireBio™ QP5355-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_006341.1 | |
Follistatin / FST Protein | |
Research Use Only | |
FST | |
Recombinant Protein | |
E. coli | |
MGNCWLRQAK . NGRCQVLYK T. ELSKEECC ST. GRLSTSW TEE. DVNDNT LFKW. MIFNG GAPNC. IPCK ETCENV. DCG PGKKCRMNKK NKPRCVC. AP DCSNITWK. G PVCGLDGKT. YRNECALLKA . RCKEQPELE V. QYQGRCKK TC. RDVFCPG SST. CVVDQT NNAY. CVTCN RICPEPASSE QYLCG. NDGV TYSSAC. HLR KATCLLG. RS IGLAYEGK. C IKAKSCEDI. QCTGGKKCLW . DFKVGRGRC S. LCDELCPD SK. SDEPVCA SDNATYASEC AMK. EAACSS GVLL. EVKHS GSCN | |
Greater than 95% as determined by SDS-PAGE. |
10468 | |
5 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Neutralization of Human Activin A induced cytotoxicity of MPC-11 cells was significantly induced by less than 200 ng/ml of Human Follistatin / FST (minimum activity of at least 5.0 x 10^3 units/mg). | |
Human | |
Untagged | |
10 mM sodium phosphate, 50mM sodium chloride, pH 7.5 |