missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fibroblast growth factor 23 Protein
A DNA sequence encoding the Human Fibroblast growth factor 23, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10385-ec-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_065689.1 | |
Fibroblast growth factor 23 Protein | |
Research Use Only | |
FGF23 | |
The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 0.5 ug/ml, corresponding to a specific activity of > 2.0 M 103 IU/mg in the presence of 0.3 ug/ml of rMuKlotho and 10 ug/ml of heparin. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
8074 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
YPNASPLLGS SWGGLIHLYT ATARNSYHLQ IHKNGHVDGA PHQTIYSALM IRSEDAGFVV ITGVMSRRYL CMDFRGNIFG SHYFDPENCR FQHQTLENGY DVYHSPQYHF LVSLGRAKRA FLPGMNPPPY SQFLSRRNEI PLIHFNTPIP RRHTRSAEDD SERDPLNVLK PRARMTPAPA SCSQELPSAE DNSPMASDPL GVVRGGRVNT HAGGTGPEGC RPFAKFI | |
Greater than 95% as determined by SDS-PAGE and HPLC. |