Learn More
enQuireBio™ Recombinant Human FGF-7 / Glrb Protein
Recombinant Protein
Marke: enQuireBio™ QP5316-500ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_445748.1 | |
FGF-7 / Glrb Protein | |
Research Use Only | |
FGF7 | |
Recombinant Protein | |
E. coli | |
MCNDMTPEQM ATNVNCSSPE RHTRSYDYME GGDIRVRRLF CRTQWYLRID KRGKVKGTQE MKNNYNIMEI RTVAVGIVAI KGVESEFYLA MNKEGKLYAK KECNEDCNFK ELILENHYNT YASAKWTHNG GEMFVALNQK GIPVRGKKTK KEQKTAHFLP MAIT | |
Greater than 90 as determined by SDS-PAGE. |
25456 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
4MBr-5 cell proliferation is significantly induced by less than 60 ng/ml of Human FGF-7 / Glrb. Activity is at least 1.7 x 10^4 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate, 100 mM soldium chloride, pH 7.5 |